Methyl-4-amino-5-bromo-2-hydroxybenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-5-bromo-2-hydroxybenzoate
056420-500MG
056420-1G
101910-444EA
698.46
USD
101910-444
101910-442
Methyl-4-amino-5-bromo-2-hydroxybenzoate ≥95%
Methyl-4-amino-5-bromo-2-hydroxybenzoate
Hazard: Irritant
Formula:
C₈H₈BrNO₃ MW: 246.07 g/mol Melting Pt: 139…141 °C |
MDL Number:
MFCD16710276 CAS Number: 286377-20-6 |
Specification Test Results
Melting point | 139-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...