6-Methoxy-2-phenylpyrimidine-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
056417-500MG
101910-432EA
1109.23
USD
101910-432
6-Methoxy-2-phenylpyrimidine-4-carboxylic acid ≥95%
6-Methoxy-2-phenylpyrimidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₀N₂O₃ Melting Pt: 153…155 °C |
MDL Number:
MFCD16710279 CAS Number: 85815-04-9 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...