6-Iodo-2,3-dihydro-1,3-benzoxazol-2-one ≥95%

Supplier: Matrix Scientific

056411-500MG 056411-1G 056411-5G
101910-404EA 629.23 USD
101910-404 101910-402 101910-400
6-Iodo-2,3-dihydro-1,3-benzoxazol-2-one ≥95%
6-Iodo-2,3-dihydro-1,3-benzoxazol-2-one

Hazard: Irritant

Formula: C₇H₄INO₂
Melting Pt: 180 °C (lit.)
MDL Number: MFCD08669499
CAS Number: 19932-87-7

Order Now

Specification Test Results

Melting point 180°C(lit)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR