N-Cyclopropyl-3-fluoro-5-iodo-4-methylbenzamide ≥95%

Supplier: Matrix Scientific

056388-500MG 056388-1G
101910-308EA 556.92 USD
101910-308 101910-310
N-Cyclopropyl-3-fluoro-5-iodo-4-methylbenzamide ≥95%
N-Cyclopropyl-3-fluoro-5-iodo-4-methylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₁FINO
Melting Pt: 117…119 °C
MDL Number: MFCD16710268
CAS Number: 585544-31-6

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR