6-Bromo-N-(2,2-dimethylpropyl)pyridine-3-carboxamide ≥95%
Supplier: Matrix Scientific
056371-500MG
056371-1G
101910-238EA
467.69
USD
101910-238
101910-236
6-Bromo-N-(2,2-dimethylpropyl)pyridine-3-carboxamide ≥95%
6-Bromo-N-(2,2-dimethylpropyl)pyridine-3-carboxamide
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂O Melting Pt: 100…102 °C |
MDL Number:
MFCD16710278 CAS Number: 1242268-19-4 |
Specification Test Results
Melting point | 100-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...