Cyclopropanesulfonamide ≥98%
Supplier: Matrix Scientific
055507-25G
055507-100G
055507-5G
101909-570EA
310.77
USD
101909-570
101909-566
101909-568
Cyclopropanesulfonamide ≥98%
Cyclopropanesulphonamide
Hazard: Irritant-Harmful
Formula:
C₃H₇NO₂S MW: 121.15 g/mol Melting Pt: 101…106 °C |
MDL Number:
MFCD08705286 CAS Number: 154350-29-5 |
Specification Test Results
Melting point | 101-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...