Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-hydroxy-3-methoxy-3,3-diphenylpropanoate

055491-500MG 055491-5G 055491-1G
101909-508EA 143.08 USD
101909-508 101909-510 101909-506
Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate ≥95%
Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate

Hazard: Irritant

Formula: C₁₇H₁₈O₄
MW: 286.33 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD09833706
CAS Number: 178306-47-3

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR