Methyl-2,4-dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylate ≥95%

Supplier: Matrix Scientific

055489-1G 055489-500MG
101909-500EA 375.38 USD
101909-500 101909-502
Methyl-2,4-dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylate ≥95%
Methyl-2,4-dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylate

Hazard: Irritant

Formula: C₁₀H₈N₂O₄
Melting Pt: 312…314 °C
MDL Number: MFCD13250068
CAS Number: 127801-84-7

Order Now

Specification Test Results

Melting point 312-314°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR