2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Piperidin-4-yl)pyridine dihydrochloride

055486-500MG
101909-490EA 763.08 USD
101909-490
2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride ≥95%
2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂
MW: 235.16 g/mol
Melting Pt: 258…261 °C
MDL Number: MFCD14583122
CAS Number: 143924-45-2

Order Now

Specification Test Results

Melting point 258-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR