2,4-Dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

055480-500MG 055480-1G
101909-466EA 747.69 USD
101909-466 101909-468
2,4-Dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylic acid ≥95%
2,4-Dioxo-1,2,3,4-tetrahydro-5-quinazolinecarboxylic acid

Hazard: Irritant

Formula: C₉H₆N₂O₄
Melting Pt: >320 °C
MDL Number: MFCD16622795
CAS Number: 5715-10-6

Order Now

Specification Test Results

Melting point 320°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR