2-[(2R,6S)-2,6-Dimethyl-1,4-oxazinan-4-yl]-5-fluorobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

055479-500MG
101909-464EA 375.38 USD
101909-464
2-[(2R,6S)-2,6-Dimethyl-1,4-oxazinan-4-yl]-5-fluorobenzenecarbaldehyde ≥95%
2-[(2R,6S)-2,6-Dimethyl-1,4-oxazinan-4-yl]-5-fluorobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₆FNO₂
Melting Pt: 91…93 °C
MDL Number: MFCD16622806
CAS Number: 679839-17-9

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR