5,8-Dichloro-7-nitroisoquinoline ≥95%

Supplier: Matrix Scientific

055478-500MG 055478-5G 055478-1G
101909-460EA 761.54 USD
101909-460 101909-462 101909-458
5,8-Dichloro-7-nitroisoquinoline ≥95%
5,8-Dichloro-7-nitroisoquinoline

Hazard: Irritant

Formula: C₉H₄Cl₂N₂O₂
Melting Pt: 190…191 °C
MDL Number: MFCD16622792
CAS Number: 1198775-29-9

Order Now

Specification Test Results

Melting point 190-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR