2-Bromo-5-methoxy-1-indanone ≥95%

Supplier: Matrix Scientific

055469-500MG 055469-1G
101909-422EA 278.46 USD
101909-422 101909-424
2-Bromo-5-methoxy-1-indanone ≥95%
2-Bromo-5-methoxy-1-indanone

Hazard: Irritant

Formula: C₁₀H₉BrO₂
Melting Pt: 106…107 °C
MDL Number: MFCD11219376
CAS Number: 29278-11-3

Order Now

Specification Test Results

Melting point 106-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR