3-(4-Bromo-2-fluorophenyl)-1H-pyrazol-5-amine ≥95%
Supplier: Matrix Scientific
055467-5G
055467-1G
055467-500MG
101909-410EA
375.38
USD
101909-410
101909-412
101909-414
3-(4-Bromo-2-fluorophenyl)-1H-pyrazol-5-amine ≥95%
3-(4-Bromo-2-fluorophenyl)-1H-pyrazol-5-amine
Hazard: Irritant
Formula:
C₉H₇BrFN₃ Melting Pt: 131…133 °C |
MDL Number:
MFCD09951350 CAS Number: 1135815-14-3 |
Specification Test Results
Melting point | 131-133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...