tert-Butyl-1,9-diazaspiro[5.5]undecane-1-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,9-Diazaspiro[5.5]undecane hydrochloride, N1-BOC protected

055460-500MG
101909-382EA 1053.85 USD
101909-382
tert-Butyl-1,9-diazaspiro[5.5]undecane-1-carboxylate hydrochloride ≥95%
tert-Butyl-1,9-diazaspiro[5.5]undecane-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₇ClN₂O₂
MW: 290.84 g/mol
Melting Pt: 180…182 °C (dec.)
MDL Number: MFCD16622777
CAS Number: 1228182-67-9

Order Now

Specification Test Results

Melting point 180-182°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR