1-[(4-Methylphenyl)sulfonyl]-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Toluene-4-sulfonyl)-1,2,3,4-tetrahydrobenzo[b]azepin-5-one1-(Toluene-4-sulphonyl)-1,2,3,4-tetrahydrobenzo[b]azepin-5-one

055451-1G 055451-5G
101909-350EA 218.46 USD
101909-350 101909-352
1-[(4-Methylphenyl)sulfonyl]-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one ≥95%
1-[(4-Methylphenyl)sulfonyl]-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one

Hazard: Irritant

Formula: C₁₇H₁₇NO₃S
MW: 315.39 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD00795994
CAS Number: 24310-36-9

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR