2-Phenethylpiperazine ≥95%

Supplier: Matrix Scientific

054914-500MG 054914-1G
101909-036EA 556.92 USD
101909-036 101909-038
2-Phenethylpiperazine ≥95%
2-Phenethylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD07373378
CAS Number: 91907-37-8

Order Now

Specification Test Results

Melting point 101-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR