5-(Methylsulfonyl)-2-thiophenecarbaldehyde ≥95%
Supplier: Matrix Scientific
054912-500MG
054912-1G
101909-030EA
356.92
USD
101909-030
101909-028
5-(Methylsulfonyl)-2-thiophenecarbaldehyde ≥95%
5-(Methylsulfonyl)-2-thiophenecarbaldehyde
Hazard: Irritant
Formula:
C₆H₆O₃S₂ Melting Pt: 108…110 °C |
MDL Number:
MFCD16140368 CAS Number: 177194-34-2 |
Specification Test Results
Melting point | 108-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...