6-(Methylsulfonyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

054907-500MG
101909-016EA 453.85 USD
101909-016
6-(Methylsulfonyl)nicotinaldehyde ≥95%
6-(Methylsulfonyl)nicotinaldehyde

Hazard: Irritant

Formula: C₇H₇NO₃S
MW: 185.2 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD16140366
CAS Number: 1221792-08-0

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR