Methyl-1,2-dimethyl-1H-1,3-benzimidazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

048707-500MG
101884-958EA 829.15 USD
101884-958
Methyl-1,2-dimethyl-1H-1,3-benzimidazole-5-carboxylate ≥95%
Methyl-1,2-dimethyl-1H-1,3-benzimidazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
Melting Pt: 161…163 °C
MDL Number: MFCD13193112
CAS Number: 256936-11-5

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR