1-Methyl-1H-1,3-benzimidazole-5-carbohydrazide ≥95%

Supplier: Matrix Scientific

048699-500MG
101884-922EA 1109.23 USD
101884-922
1-Methyl-1H-1,3-benzimidazole-5-carbohydrazide ≥95%
1-Methyl-1H-1,3-benzimidazole-5-carbohydrazide

Hazard: Irritant

Formula: C₉H₁₀N₄O
Melting Pt: 250…252 °C
MDL Number: MFCD13193107
CAS Number: 1192263-92-5

Order Now

Specification Test Results

Melting point 250-252°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR