Methyl-4-aminopicolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Aminopicolinic acid methyll ester
, Methyl 4-Aminopicolinate
, Methyl 4-Aminopyridine-2-carboxylate
, 4-Aminopyridine-2-carboxylic acid methyll ester
048697-1G
101884-916EA
55.38
USD
101884-916
Methyl-4-aminopicolinate ≥95%
Methyl 4-aminopicolinate
Hazard: Irritant
Formula:
C₇H₈N₂O₂ MW: 152.1500 g/mol Melting Pt: 129…130 °C |
MDL Number:
MFCD00956000 CAS Number: 71469-93-7 |
Specification Test Results
Melting point | 129-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...