Ethyl-5-bromo-4,6-dichloronicotinate ≥95%
Supplier: Matrix Scientific
048688-1G
048688-500MG
101884-878EA
681.54
USD
101884-878
101884-876
Ethyl-5-bromo-4,6-dichloronicotinate ≥95%
Ethyl-5-bromo-4,6-dichloronicotinate
Hazard: Irritant
Formula:
C₈H₆BrCl₂NO₂ MW: 298.96 g/mol Melting Pt: 35…37 °C |
MDL Number:
MFCD13193100 CAS Number: 1192263-86-7 |
Specification Test Results
Melting point | 35-37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...