3,4-Dinitrosobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

048686-500MG
101884-870EA 747.69 USD
101884-870
3,4-Dinitrosobenzenecarbonitrile ≥95%
3,4-Dinitrosobenzenecarbonitrile

Hazard: Irritant

Formula: C₇H₃N₃O₂
Melting Pt: 75…77 °C
MDL Number: MFCD13193098
CAS Number: 1192263-84-5

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR