3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-2H-pyrido[3,2-b]-1,4-oxazine

048685-500MG 048685-1G 048685-5G
101884-864EA 100 USD
101884-864 101884-868 101884-866
3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine ≥95%
3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD09025907
CAS Number: 20348-23-6

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR