1H-Pyrazolo[3,4-b]pyridin-3(2H)-one ≥95%

Supplier: Matrix Scientific

048684-500MG
101884-862EA 338.46 USD
101884-862
1H-Pyrazolo[3,4-b]pyridin-3(2H)-one ≥95%
1H-Pyrazolo[3,4-b]pyridin-3(2H)-one

Hazard: Irritant

Formula: C₆H₅N₃O
MW: 135.12 g/mol
Melting Pt: 234…236 °C
MDL Number: MFCD03013435
CAS Number: 2942-43-0

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR