4,6-Dichloronicotinamide ≥97%

Supplier: Matrix Scientific

048681-1G
101884-852EA 167.69 USD
101884-852
4,6-Dichloronicotinamide ≥97%
4,6-Dichloronicotinamide

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Melting Pt: 153…155 °C
MDL Number: MFCD01646073
CAS Number: 70593-57-6

Order Now

Specification Test Results

Melting point 153-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR