4-[(Dibenzylamino)methyl]-4-piperidinol ≥95%

Supplier: Matrix Scientific

048677-500MG
101884-836EA 688.85 USD
101884-836
4-[(Dibenzylamino)methyl]-4-piperidinol ≥95%
4-[(Dibenzylamino)methyl]-4-piperidinol

Hazard: Irritant

Formula: C₂₀H₂₆N₂O
Melting Pt: 132…140 °C
MDL Number: MFCD13193094
CAS Number: 1192263-91-4

Order Now

Specification Test Results

Melting point 132-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR