4-Chloro-7-nitro-1H-indazol-3-amine ≥95%
Supplier: Matrix Scientific
048676-1G
048676-500MG
101884-834EA
467.69
USD
101884-834
101884-832
4-Chloro-7-nitro-1H-indazol-3-amine ≥95%
4-Chloro-7-nitro-1H-indazol-3-amine
Hazard: Irritant
Formula:
C₇H₅ClN₄O₂ Melting Pt: 234…236 °C |
MDL Number:
MFCD13193093 CAS Number: 1192263-96-9 |
Specification Test Results
Melting point | 234-236°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...