2-Chloro-5-iodo-4-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-chloro-5-iodopyridine

048673-250MG 048673-5G 048673-1G
101884-820EA 106.15 USD
101884-820 101884-822 101884-818
2-Chloro-5-iodo-4-pyridinamine ≥95%
2-Chloro-5-iodo-4-pyridinamine

Hazard: Irritant-Harmful

Formula: C₅H₄ClIN₂
MW: 254.455 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD09834661
CAS Number: 800402-12-4

Order Now

Specification Test Results

Melting point 126-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR