3-Bromo-2-chloro-5-methyl[1,6]naphthyridine ≥95%

Supplier: Matrix Scientific

048655-500MG
101884-750EA 1023.5 USD
101884-750
3-Bromo-2-chloro-5-methyl[1,6]naphthyridine ≥95%
3-Bromo-2-chloro-5-methyl[1,6]naphthyridine

Hazard: Irritant

Formula: C₉H₆BrClN₂
Melting Pt: 123…125 °C
MDL Number: MFCD13193079
CAS Number: 1192263-93-6

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR