2-Amino-4,6-bis(trifluoromethyl)nicotinamide ≥95%

Supplier: Matrix Scientific

048646-500MG 048646-1G
101884-714EA 556.92 USD
101884-714 101884-716
2-Amino-4,6-bis(trifluoromethyl)nicotinamide ≥95%
2-Amino-4,6-bis(trifluoromethyl)nicotinamide

Hazard: Irritant

Formula: C₈H₅F₆N₃O
Melting Pt: 188…190 °C
MDL Number: MFCD13193070
CAS Number: 89990-38-5

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR