tert-Butyl-2-(4-aminobenzoyl)-1-pyrazolidinecarboxylate ≥95%

Supplier: Matrix Scientific

048645-1G 048645-500MG
101884-710EA 688.85 USD
101884-710 101884-712
tert-Butyl-2-(4-aminobenzoyl)-1-pyrazolidinecarboxylate ≥95%
tert-Butyl-2-(4-aminobenzoyl)-1-pyrazolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁N₃O₃
Melting Pt: 137…140 °C
MDL Number: MFCD13193069
CAS Number: 1198475-42-1

Order Now

Specification Test Results

Melting point 137-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR