2-Methyl-5-(trifluoromethyl)-1,3-benzothiazole-6-carbonitrile ≥95%

Supplier: Matrix Scientific

048634-500MG
101884-672EA 829.15 USD
101884-672
2-Methyl-5-(trifluoromethyl)-1,3-benzothiazole-6-carbonitrile ≥95%
2-Methyl-5-(trifluoromethyl)-1,3-benzothiazole-6-carbonitrile

Hazard: Irritant

Formula: C₁₀H₅F₃N₂S
Melting Pt: 106…108 °C
MDL Number: MFCD12922695
CAS Number: 1190198-32-3

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR