Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
048630-1G
048630-500MG
101884-654EA
669.23
USD
101884-654
101884-656
Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate ≥95%
Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₇H₅F₃N₂O₄ MW: 238.12 g/mol Melting Pt: 225…226 °C |
MDL Number:
MFCD12922685 CAS Number: 936476-63-0 |
Specification Test Results
Melting point | 225-226°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...