Methyl-4,6-dihydroxy-5-iodonicotinate ≥95%

Supplier: Matrix Scientific

048628-1G 048628-500MG
101884-644EA 280.6 USD
101884-644 101884-646
Methyl-4,6-dihydroxy-5-iodonicotinate ≥95%
Methyl-4,6-dihydroxy-5-iodonicotinate

Hazard: Irritant

Formula: C₇H₆INO₄
MW: 295.03 g/mol
Melting Pt: 255…257 °C
MDL Number: MFCD12922714
CAS Number: 1190198-28-7

Order Now

Specification Test Results

Melting point 255-257°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR