2,3-Dichloropyridin-4-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2,3-dichloropyridine
048601-1G
048601-500MG
101884-542EA
460
USD
101884-542
101884-540
2,3-Dichloropyridin-4-amine ≥95%
2,3-Dichloropyridin-4-amine
Hazard: Irritant
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Melting Pt: 152…153 °C |
MDL Number:
MFCD01646113 CAS Number: 184416-83-9 |
Specification Test Results
Melting point | 152-153°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...