2-Bromo-4-nitro-5-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

048583-500MG 048583-1G
101884-468EA 438.46 USD
101884-468 101884-466
2-Bromo-4-nitro-5-(trifluoromethyl)aniline ≥95%
2-Bromo-4-nitro-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄BrF₃N₂O₂
MW: 285.0181 g/mol
Melting Pt: 113…114 °C
MDL Number: MFCD12922693
CAS Number: 1190198-31-2

Order Now

Specification Test Results

Melting point 113-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR