5-Bromo-2-methoxy-6-(methylamino)nicotinic acid ≥95%

Supplier: Matrix Scientific

048580-500MG
101884-456EA 1296.92 USD
101884-456
5-Bromo-2-methoxy-6-(methylamino)nicotinic acid ≥95%
5-Bromo-2-methoxy-6-(methylamino)nicotinic acid

Hazard: Irritant

Formula: C₈H₉BrN₂O₃
Melting Pt: 207…209 °C
MDL Number: MFCD12922708
CAS Number: 187480-17-7

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR