5-(Benzyloxy)-2-(chloromethyl)-4-pyridinol ≥95%
Supplier: Matrix Scientific
048571-1G
048571-500MG
101884-428EA
698.46
USD
101884-428
101884-426
5-(Benzyloxy)-2-(chloromethyl)-4-pyridinol ≥95%
5-(Benzyloxy)-2-(chloromethyl)-4-pyridinol
Hazard: Irritant
Formula:
C₁₃H₁₂ClNO₂ Melting Pt: 178…179 °C |
MDL Number:
MFCD12922672 CAS Number: 1105193-30-3 |
Specification Test Results
Melting point | 178-179°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...