2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5,7-dihydro-4H-thieno[2,3-c]pyran-3-carboxamide

048566-500MG 048566-1G
101884-402EA 363.08 USD
101884-402 101884-404
2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxamide ≥95%
2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxamide

Hazard: Irritant

Formula: C₈H₁₀N₂O₂S
MW: 198.25 g/mol
Melting Pt: 222…223 °C
MDL Number: MFCD12087875
CAS Number: 848324-33-4

Order Now

Specification Test Results

Melting point 222-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR