3-Oxo-3-thieno[2,3-b]pyridin-2-ylpropanenitrile ≥95%

Supplier: Matrix Scientific

048557-500MG 048557-1G
101884-366EA 698.46 USD
101884-366 101884-364
3-Oxo-3-thieno[2,3-b]pyridin-2-ylpropanenitrile ≥95%
3-Oxo-3-thieno[2,3-b]pyridin-2-ylpropanenitrile

Hazard: Irritant

Formula: C₁₀H₆N₂OS
Melting Pt: 204…206 °C
MDL Number: MFCD12827817
CAS Number: 1186405-15-1

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR