Methyl-2-methoxy-6-(methylamino)nicotinate ≥95%

Supplier: Matrix Scientific

048546-500MG
101884-318EA 564.65 USD
101884-318
Methyl-2-methoxy-6-(methylamino)nicotinate ≥95%
Methyl-2-methoxy-6-(methylamino)nicotinate

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MW: 196.21 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD12827839
CAS Number: 187480-13-3

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR