6-Iodo-4-oxo-1,4-dihydro-3-quinoline-carboxylic acid ≥95%

Supplier: Matrix Scientific

048538-1G 048538-500MG 048538-5G
101884-278EA 375.38 USD
101884-278 101884-280 101884-282
6-Iodo-4-oxo-1,4-dihydro-3-quinoline-carboxylic acid ≥95%
6-Iodo-4-oxo-1,4-dihydro-3-quinoline-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆INO₃
Melting Pt: 295 °C (dec.)
MDL Number: MFCD01912285
CAS Number: 40107-06-0

Order Now

Specification Test Results

Melting point 295(Dec)°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR