Ethyl-4,6-dichloro-5-nitronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,6-dichloro-5-nitronicotinate

048530-500MG 048530-1G
101884-238EA 336.92 USD
101884-238 101884-240
Ethyl-4,6-dichloro-5-nitronicotinate ≥95%
Ethyl-4,6-dichloro-5-nitronicotinate

Hazard: Irritant

Formula: C₈H₆Cl₂N₂O₄
MW: 265.05 g/mol
Melting Pt: 36…38 °C
MDL Number: MFCD09842620
CAS Number: 154012-15-4

Order Now

Specification Test Results

Melting point 36-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR