2-Bromo-1,3-cyclopentanedione ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromocyclopentane-1,3-dione

048508-500MG
101884-136EA 335.8 USD
101884-136
2-Bromo-1,3-cyclopentanedione ≥95%
2-Bromo-1,3-cyclopentanedione

Hazard: Irritant

Formula: C₅H₅BrO₂
MW: 177 g/mol
Melting Pt: 197…199 °C (dec.)
MDL Number: MFCD12827833
CAS Number: 14203-24-8

Order Now

Specification Test Results

Melting point 197-199DEC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR