2-(Benzylamino)-5-chloronicotinic acid ≥95%

Supplier: Matrix Scientific

048501-500MG
101884-106EA 280.6 USD
101884-106
2-(Benzylamino)-5-chloronicotinic acid ≥95%
2-(Benzylamino)-5-chloronicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O₂
Melting Pt: 228…230 °C
MDL Number: MFCD12827795
CAS Number: 750549-62-3

Order Now

Specification Test Results

Melting point 228-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR