3-Azabicyclo[3.2.0]heptane-2,4-dione ≥95%
Supplier: Matrix Scientific
048500-500MG
101884-104EA
747.69
USD
101884-104
3-Azabicyclo[3.2.0]heptane-2,4-dione ≥95%
3-Azabicyclo[3.2.0]heptane-2,4-dione
Hazard: Irritant
Formula:
C₆H₇NO₂ Melting Pt: 134…135 °C |
MDL Number:
MFCD12827834 CAS Number: 1122-09-4 |
Specification Test Results
Melting point | 134-135°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...