Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

048216-1G 048216-500MG
101883-330EA 416.3 USD
101883-330 101883-332
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate ≥95%
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD11841051
CAS Number: 1135283-79-2

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR