Methyl-5-(dimethylamino)-2-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

048211-500MG
101883-312EA 280.6 USD
101883-312
Methyl-5-(dimethylamino)-2-nitrobenzenecarboxylate ≥95%
Methyl-5-(dimethylamino)-2-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
Melting Pt: 98…100 °C
MDL Number: MFCD11841048
CAS Number: 749863-29-4

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR